Learn More
Medchemexpress LLC HY-141513 5mg , NH-3 CAS:447415-26-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1415135MG
Encompass_Preferred
HY-141513 5mg NH-3 CAS: 447415-26-1 NH-3 is an orally active, reversible thyroid hormone receptor (THR) antagonist with an IC50 of 55 nM. NH-3, a derivative of the selective thyromi-metic GC-1, inhibits binding of thyroid hormones to their receptor and that inhibits cofactor recruitment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0846