Learn More
Medchemexpress LLC NH-bis(C2-PEG2-NH-Boc) | 2182601-69-8 | 50 MG

Supplier: Medchemexpress LLC HY13053150MG
NH-bis(C2-PEG2-NH-Boc) is a laboratory chemical identified for use in the manufacture of substances. This compound is a stable liquid with a molecular formula of C22H45N3O8 and a molecular weight of 479.61.
- Identified for use as a laboratory chemical
- Stable under recommended storage conditions
- Pure form storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Shipping at room temperature if less than 2 weeks
- Appearance: liquid
- Boiling point/range: 577.0±50.0 °C at 760 mmHg
- Relative density: 1.064 g/cm³
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.