Learn More
Medchemexpress LLC NH-bis(PEG3-azide) | 1258939-39-7 | 98.0% | C16H33N7O6 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140551
Encompass_Preferred
NH-bis(PEG3-azide) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Azide group, and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Capable of strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-52720