Learn More
Medchemexpress LLC NH-bis-PEG4 | 63721-14-2 | 98.0% | C16H35NO8 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1405481G
Encompass_Preferred
NH-bis-PEG4 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is intended for research use only. It is designed to facilitate the targeted degradation of proteins by exploiting the ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs
  • PEG-based linker design
  • Facilitates selective degradation of target proteins
  • Intended for research applications
Catalog No. 50-003-56228