Learn More
Medchemexpress LLC NH-bis(PEG4-Boc) | 2055041-41-1 | 98.0% | C30H59NO12 | 25 MG

Supplier: Medchemexpress LLC 25MGHY140258
NH-bis(PEG4-Boc) is a chemical compound with the molecular formula C30H59NO12 and a molecular weight of 625.79. It is typically a colorless to light yellow liquid with a purity of 98.0% as determined by NMR.
- Appearance: Colorless to light yellow liquid
- Molecular weight: 625.79
- Purity (NMR): 98.0%
- Storage conditions: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.