Learn More
Medchemexpress LLC NH-bis(PEG4-Boc) | 2055041-41-1 | 98.0% | C30H59NO12 | 50 MG

Supplier: Medchemexpress LLC HY14025850MG
NH-bis(PEG4-Boc) is a chemical compound primarily intended for research use. It is identified as 4,7,10,13,19,22,25,28-Octaoxa-16-azahentriacontanedioic acid, 1,31-bis(1,1-dimethylethyl) ester.
- Purity (NMR): 98.0%
- Molecular formula: C30H59NO12
- Molecular weight: 625.79
- Appearance: Colorless to light yellow liquid
- Recommended storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.