Learn More
Broadpharm NH-bis(PEG4-Boc) | 2182601-75-6 | MFCD31536762 | 500mg
Supplier:  Broadpharm BP23782500MG
Encompass
NH-bis(PEG4-Boc) | Putity: 98% | MW: 655.8 | 2182601-75-6 | MFCD31536762 | 500mg
Catalog No. NC1657179