Learn More
Medchemexpress LLC NH-bis(PEG4-C2-NH-Boc) | 2182601-75-6 | MFCD31536762 | ≥97.0% | C30H61N3O12 | 25 MG

Supplier: Medchemexpress LLC 25MGHY140261
NH-bis(PEG4-C2-NH-Boc) is a colorless to light yellow liquid. This PEG-based PROTAC linker is used in the synthesis of PROTACs for research applications.
- Cas number: 2182601-75-6
- Chemical formula: C30H61N3O12
- Purity: ≥97.0%
- Appearance: Colorless to light yellow liquid
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.