Learn More
Medchemexpress LLC NH2-C5-NH-Boc | 51644-96-3 | 202.29 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0047105G
Encompass_Preferred
NH2-C5-NH-Boc (PROTAC Linker 23) is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and not sold to patients.
  • Alkyl chain-based linker
  • Used in the synthesis of protacs
  • For research use only
Catalog No. 50-004-49069