Learn More
Medchemexpress LLC NH2-C5-PEG4-N3-L-lysine-PEG3-N3 | 99.4% | 619.71 Da | C25H49N9O9 | 25 MG

Supplier: Medchemexpress LLC HY13094625MG
NH2-C5-PEG4-N3-L-lysine-PEG3-N3 is a cleavable polyethylene glycol (PEG) based linker designed for antibody-drug conjugation and bioconjugation chemistry. It supplies an amino terminus and azide groups for conjugation, and is provided as a high-purity, water-soluble reagent.
- Cleavable 7-unit PEG linker suitable for ADC synthesis.
- Contains amino and azide functional groups for bioconjugation.
- High purity, suitable for research applications.
- Water soluble (≥100 mg/mL), enabling aqueous handling.
- Molecular weight approximately 619.71 Da for predictable stoichiometry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.