Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-C5-PEG4-N3-L-lysine-PEG3-N3 | 99.4% | 619.71 g/mol | C25H49N9O9 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1309461MG

Encompass_Preferred

NH2-C5-PEG4-N3-L-lysine-PEG3-N3 is a cleavable 7-unit polyethylene glycol (PEG) linker bearing an azide functionality for use in click chemistry and antibody-drug conjugate (ADC) synthesis. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) bioconjugation workflows, and is supplied as a highly pure, water-soluble reagent.

  • Cleavable 7-unit PEG linker
  • Contains an azide group for click chemistry
  • Suitable for CuAAC and SPAAC bioconjugation
  • Soluble in water at ≥ 100 mg/mL
  • High purity: 99.42%
  • Available in small-scale quantities for research use

Catalog No. 50-002-04141


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.