Learn More
YMC America, Inc NH2 col 120A 3um 100x3mm
SDP
Supplier:  YMC America, Inc NH12S031003WT
Encompass_Preferred
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-78473