Learn More
Medchemexpress LLC NH2 DOTA-GA | 1639843-65-4 | 100.0% | 518.56 g·mol⁻¹ | C21H38N6O9 | 25 MG

Supplier: Medchemexpress LLC HY16456925MG
NH2 DOTA-GA is a DOTA-based bifunctional metal chelator used to prepare radionuclide drug conjugates. It is supplied as a solid powder and contains carboxylic acid groups capable of complexing metal ions, such as Gd(III).
- DOTA-based bifunctional chelator for radionuclide conjugates.
- Useful for metal ion complexation, including Gd(III).
- CAS number 1639843-65-4 for unambiguous identification.
- Molecular formula C21H38N6O9 and molecular weight 518.56 g·mol⁻¹.
- High chemical purity suitable for research applications.
- Solid powder with recommended storage at -20°C (powder) or -80°C/-20°C in solvent.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.