Learn More
Medchemexpress LLC NH2-Gly-PAB-Exatecan-D-glucuronic acid (formic) | 99.3% | 879.79 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY160885A
Encompass_Preferred
NH2-Gly-PAB-Exatecan-D-glucuronic acid formic (Compound 8) is the formic salt form of H2N-Gly-PAB-Exatecan-D-glucuronic acid. It is an intermediate that reacts with an NHS group to form a drug-linker conjugate for ADC.
  • Intermediate used in the formation of drug-linker conjugates
  • Target: ADC payload
  • Pathway: Antibody-drug conjugate/ADC related
Catalog No. 50-003-92497