Learn More
YMC America, Inc NH2 guard 120A 3um 10x2.1mm
SDP
Supplier:  YMC America, Inc NH12S0301Q1GC
Encompass
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-78754