Learn More
Medchemexpress LLC NH2-PEG2-CH2-Boc | 1948273-09-3 | ≥97.0% | 233.30 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY42427
Encompass_Preferred
NH2-PEG2-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-02408