Learn More
Medchemexpress LLC NH2-PEG8-OH | 352439-37-3 | 97.0% | 250 MG

Supplier: Medchemexpress LLC 250MGHY130200
NH2-PEG8-OH is a PEG-based PROTAC linker primarily used for laboratory research and the manufacture of various substances. This chemical compound is identified by its CAS number 352439-37-3.
- Peg-based protac linker
- Suitable for laboratory research
- Utilized in the manufacture of substances
- Molecular formula: C16H35NO8
- Molecular weight: 369.45
- Appearance: Solid-liquid mixture
- Purity: 97.0%
- Recommended storage at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.