Learn More
Lonza Walkersville NHBE – Human Bronchial/Tracheal Epithelial Cells without Retinoic Acid
Supplier:  Lonza Walkersville CC2541
Encompass
Normal and Diseased Bronchial Epithelial (NHBE and DHBE) cells are isolated from epithelial lining of airways above bifurcation of the lungs. Lonza's Normal Human Bronchial Epithelial Cells (NHBE) without retinoic acid are available. Certain lots with Small Airway epithelial cells (CC-2547) or normal human lung fibroblasts (CC-2512)from matched donors are available.Cryopreserved human bronchial epithelial cells without retinoic acid are guaranteed through 15 population doublings. Cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Content:1 x Cryopreserved ampule containing ≥500,000 cells
Catalog No. NC2398705