Learn More
Lonza Walkersville NHCF-V – Human Ventricular Cardiac Fibroblasts
Supplier:  Lonza Walkersville CC2904
Encompass
Human Cardiac-Related Fibroblasts include Aortic Adventitial Fibroblasts (AoAF) isolated from the tunica external of the ascending or descending aorta and cardiac fibroblasts (NHCF-A and -V) isolated from the atrium or ventricle. Cells are isolated from normal, adult heart tissue. Cryopreserved cells are shipped at passage 2. Benefits:Assured for experimental use up to 15 population doublings Express ≥90% collagen I and ≤10% Von Willibrand factor VIIIAll cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lotApplications:Vascular diseasesNeointimal formationVascular remodelingAtherosclerosisHypertensionMyocardial repairExtracellular maintenance during cardiac cell / tissue growthCardiac Fibrosis Paracrine signallingToxicologyGene expressionContent:1 x Cryopreserved ampule NHCF-V containing ≥ 500,000 cells/vial
Catalog No. NC1763063