Learn More
Promocell GmbH PromoCell NHDF-c 500,000 Human Dermal Fibroblasts, cryopreserved
Supplier:  Promocell GmbH C12300
Encompass
Primary Normal Human Dermal Fibroblasts (NHDF) are isolated form the dermis of juvenile foreskin or adult skin from different locations like the face, the breasts, the abdomen, and the thighs.They can be used for wound healing studies and dermatological research to investigate diseases like scleroderma, fibrosarcoma, fibrosis, xeroderma pigmentosum, and histiocytoma. Moreover, fibroblasts are important for cancer research, tissue regeneration, and tissue engineering studies.
Catalog No. 50-306-002