Learn More
ABclonal Technology NHLH1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A24828100UL
Encompass_Preferred
The helix-loop-helix (HLH) proteins are a family of putative transcription factors some of which have been shown to play an important role in growth and development of a wide variety of tissues and species Four members of this family have been clearly implicated in tumorigenesis via their involvement in chromosomal translocations in lymphoid tumors MYC (MIM 190080) LYL1 (MIM 151440) E2A (MIM 147141) and SCL (MIM 187040)
Catalog No. 50-000-09860