Learn More
Medchemexpress LLC Nhpi-PEG4-C2-NHS ester | 1415328-95-8 | 98.9% | 508.48 g/mol | C23H28N2O11 | 250 MG

Supplier: Medchemexpress LLC HY130091250MG
NHPI-PEG4-C2-NHS ester is an NHPI-activated PEG4-based NHS ester linker used for bioconjugation and antibody-drug conjugate (ADC) synthesis. It is supplied as a solid for research use and is reported at approximately 98.9% purity.
- CAS number: 1415328-95-8.
- Molecular weight 508.48 g/mol.
- Chemical formula C23H28N2O11.
- Useful as an NHS ester for amine coupling and ADC linker synthesis.
- Available in multiple package sizes, including 250 MG.
- For research use only; not for clinical or human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.