Learn More
Medchemexpress LLC NHS-MMAF | 1404073-19-3 | 97.2% | 943.18 | 25 MG

Supplier: Medchemexpress LLC 25MGHY139326
NHS-MMAF is a modified MMAF (Monomethyl auristatin F) extracted from patent WO2012143499. It is a potent tubulin polymerization inhibitor and is utilized as an antitumor agent. This product is intended for research use only and is not for sale to patients.
- Potent tubulin polymerization inhibitor
- Antitumor agent
- Research use only
- Soluble in DMSO
- Store at -80°C for up to 6 months or -20°C for up to 1 month (sealed, away from moisture and light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.