Learn More
Medchemexpress LLC NHS-MMAF | 1404073-19-3 | 97.2% | C49H78N6O12 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13932610MG
Encompass_Preferred
NHS-MMAF is an N-hydroxysuccinimide-activated derivative of monomethyl auristatin F used as a payload intermediate for antibody-drug conjugate (ADC) synthesis. It acts as a tubulin polymerization inhibitor and is intended for research use in ADC linker and conjugation development; it is described in patent WO2012143499 (intermediate 219).
  • NHS-activated ester for amine conjugation.
  • Suitable for antibody-drug conjugate linker chemistry.
  • High reported purity (97.2%).
  • Solid, white to off-white appearance for easy handling.
  • Soluble in DMSO for reaction setup.
  • Serves as a payload precursor for cytotoxic conjugates.
Catalog No. 50-002-59621