Learn More
Medchemexpress LLC Nhs-peg2-ss-peg2-nhs | 2640393-71-9 | 96.7% | 580.63 | C22H32N2O12S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY13613310MG
Encompass_Preferred
NHS-PEG2-SS-PEG2-NHS is a cleavable four-unit polyethylene glycol (PEG) linker bearing N-hydroxysuccinimide (NHS) esters at both termini. It is designed for use in bioconjugation workflows, including the synthesis of antibody-drug conjugates (ADCs), providing a reducible disulfide-containing spacer to improve solubility and enable payload release under reducing conditions.
  • Cleavable disulfide-containing PEG spacer for controlled payload release.
  • Terminal NHS esters for efficient conjugation to amines.
  • Four-unit PEG improves aqueous solubility of conjugates.
  • Suitable for ADC synthesis and general bioconjugation workflows.
  • Intended for research use; handle according to safety data sheet instructions.
Catalog No. 50-002-03867