Learn More
Medchemexpress LLC Nhs-PEG2-SS-PEG2-NHS | 580.63 | C22H32N2O12S2 | 50 MG

Supplier: Medchemexpress LLC HY13613350MG
NHS-PEG2-SS-PEG2-NHS is a cleavable, homobifunctional polyethylene glycol (PEG) linker containing a central disulfide bond and two N-hydroxysuccinimide (NHS) activated esters. It is intended for bioconjugation and use as an antibody-drug conjugate (ADC) linker in research settings.
- Molecular formula: C22H32N2O12S2.
- Molecular weight: 580.63.
- Contains a reducible disulfide (cleavable) linker between PEG2 units.
- Homobifunctional with two NHS activated esters for amine coupling.
- Supplied in small laboratory quantity suitable for research use.
- Storage: store sealed at -20°C, protect from moisture; in solution, recommended storage at -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.