Learn More
Medchemexpress LLC NHS-SS-biotin | 122266-55-1 | MFCD28505564 | 98.0% | 504.64 g/mol | C19H28N4O6S3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140129100MG
Encompass_Preferred
NHS-SS-biotin is a cleavable disulfide-containing biotin linker with an N-hydroxysuccinimide (NHS) ester reactive group. It is used to introduce a biotin tag onto primary amines and as a reducible linker in antibody-drug conjugate synthesis and other bioconjugation workflows.
  • Cleavable disulfide linker for reductive release.
  • NHS ester reactive toward primary amines.
  • Useful for protein and peptide biotinylation and affinity capture.
  • Available in small-scale quantities suitable for research applications.
  • Purity typically 98.0% as provided in supplier documentation.
Catalog No. 50-002-03976