Learn More
Sigma Aldrich Fine Chemicals Biosciences Ni SepharoseR High Perform
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17526801
Encompass
Ni Sepharose(R) High Performance affinity media for high-reSOLUTIONon high capacity purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).Ni Sepharose(R) High Performance consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with Ni2 which will selectively retain proteins with exposed histidine groups.The high stability and broad compatibility of Ni Sepharose(R) High Performance maintains biological activity and increases product yield at the same time as it greatly expands the range of suitable operating conditions.
Catalog No. 50-290-4092