Learn More
Medchemexpress LLC Niacin (standard) | 59-67-6 | 99.96% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB014310G
Encompass_Preferred
Niacin (Vitamin B3) is an orally active water-soluble B3 vitamin and an essential nutrient for humans. It plays a key role in energy metabolism and cell signaling cascades regulating gene expression and apoptosis. This product is intended for research and analytical applications, and is also used in the study of cardiovascular diseases.
  • Orally active water-soluble B3 vitamin
  • Essential nutrient for humans
  • Plays a key role in energy metabolism
  • Regulates gene expression and apoptosis through cell signaling cascades
  • Used in the study of cardiovascular diseases
  • Intended for research and analytical applications
Catalog No. 50-004-50897