Learn More
Medchemexpress LLC Nialamide | 51-12-7 | 96.3% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB119910MM/1ML
Encompass_Preferred
Nialamide is a non-selective monoamine oxidase (MAO) inhibitor that inhibits MAO and regulates reactive oxygen species (ROS) production. It can be used in the research of depression, inflammatory diseases, neurodegenerative diseases, and hypertension.
  • Inhibits MAO
  • Regulates reactive oxygen species (ROS) production
  • Induces hyperkinesis in animals
  • Enhances anticonvulsant effect
  • Increases rectal temperature
  • Enhances pressor effect of Norepinephrine
  • Suitable for research of depression
  • Suitable for research of inflammatory diseases
  • Suitable for research of neurodegenerative diseases
  • Suitable for research of hypertension
Catalog No. 50-004-11289