Learn More
Medchemexpress LLC Niaprazine | 27367-90-4 | MFCD00867991 | 99.6% | 356.44 g/mol | C20H25FN4O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY105542R100MG
Encompass_Preferred
Niaprazine (Standard) is an analytical reference material of niaprazine intended for research and analytical applications. It acts as a histamine H1-receptor antagonist with antihistamine and antiserotonin activities and is commonly used in pharmacological and sleep disorder studies. The product is supplied as a 100 mg solid with documented purity and supporting analytical documentation.
  • Analytical reference standard for research and method validation.
  • High purity: 99.6%.
  • Molecular formula C20H25FN4O; molecular weight 356.44 g/mol.
  • Suitable for pharmacological and sleep disorder research.
  • Includes supporting documentation such as SDS and COA.
Catalog No. 50-002-42054