Learn More
Medchemexpress LLC HY-105542 5mg Medchemexpress, Niaprazine CAS:27367-90-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1055425MG
Encompass_Preferred
Medchemexpress, HY-105542 5mg Niaprazine CAS:27367-90-4 Niaprazine is a histamine H1-receptor antagonist with marked sedative properties. Niaprazine has antihistamine and antiserotonin activities and can be used for sleep disorder research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0685