Learn More
Medchemexpress LLC NIC-0102 | 2806031-94-5 | C21H25BF2N2O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15125210MM/1ML
Encompass_Preferred
NIC-0102 is an orally active proteasome inhibitor that specifically inhibits NLRP3 inflammatory vesicle activation. It demonstrates potent anti-inflammatory effects in a model of dextran sulfate sodium (DSS)-induced ulcerative colitis.
  • Inhibits proteasome with a pIC50 of 7.55
  • Specifically inhibits NLRP3 inflammatory vesicle activation
  • Shows potent anti-inflammatory effects
  • Inhibits production of pro-IL-1β
Catalog No. 50-003-75892