Learn More
Medchemexpress LLC Nicardipine-d3 hydrochloride | 1432061-50-1 | 99.7% | 519.00 | C26H27D3ClN3O6 | 5MG
SDP
Supplier:  Medchemexpress LLC HY12515AS5MG
Encompass_Preferred
This deuterium-labeled hydrochloride salt of nicardipine is supplied at high purity as a small-quantity analytical reference for mass spectrometry, method development, and pharmacological research. It provides a stable isotopic standard for accurate quantitation and method verification in analytical workflows.
  • High isotopic labeling for accurate mass spectrometry internal standard use.
  • High chemical purity suitable for analytical applications.
  • Hydrochloride salt form for improved solubility in common solvents.
  • Supplied in a small quantity suitable for method development and reference use.
  • Appropriate as a reference material for pharmacology and analytical chemistry workflows.
Catalog No. 50-002-38655