Learn More
Medchemexpress LLC Nicardipine hydrochloride | 54527-84-3 | MFCD00057327 | 99.73% | 515.99 g/mol | C26H30ClN3O6 | 1G

Supplier: Medchemexpress LLC HY12515A1G
Nicardipine hydrochloride is the hydrochloride salt of nicardipine, a dihydropyridine L-type calcium channel blocker supplied for research and analytical use. It is commonly used in studies of cardiovascular pharmacology, including antihypertensive and antianginal research. CAS 54527-84-3; purity 99.73%.
- High purity suitable for analytical and research applications.
- Acts as an L-type calcium channel blocker used in cardiovascular research.
- Hydrochloride salt form for improved solubility in aqueous media.
- CAS number 54527-84-3 for unambiguous identification.
- Molecular formula C26H30ClN3O6; molecular weight ~516.0 g/mol.
- Supplied with supporting documentation such as a datasheet or certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.