Learn More
Cayman Chemical NIcardIpInhydrochlorIde 1g
SDP
Supplier:  Cayman Chemical 175371 G
Encompass_Preferred
A dihydropyridine L-type calcium channel antagonist that displays antihypertensive and antianginal activity; selectively inhibits adenosine A3 receptors (Ki = 3.25 µM) and CYP3A4 catalytic activity (IC50 = 0.148 µM); activates TRPA1 channels, producing an increase in Ca2+ (EC50 = 0.5 µM)
Catalog No. 50-296-7920