Learn More
Medchemexpress LLC Nicastrin protein, human (HEK293, His) | >95.0% | 100 UG

Supplier: Medchemexpress LLC HYP74698100UG
Recombinant human Nicastrin (residues N34-E669) expressed in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized protein for research use, it is suitable for biochemical and cell-based studies of the gamma-secretase complex, protein-protein interactions, and assay development.
- Expression system: HEK293 cells.
- Tag: C-terminal His tag.
- Sequence: residues N34-E669.
- Purity: >95.0% by reducing SDS-PAGE.
- Format and quantity: lyophilized powder, 100 μg.
- Storage and handling: store lyophilized at -20°C; reconstitute to ≥100 μg/mL and aliquot to avoid freeze-thaw.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.