Learn More
Strem, An Ascensus Company CAS# 39430-27-8. 50g. Nickel(II) carbonate, basic hydrate. MFCD00150262
SDP
Supplier:  Strem, An Ascensus Company 93280850G
Encompass
CAS# 39430-27-8. 50g. Nickel(II) carbonate, basic hydrate. MFCD00150262. Molecular Weight: 320.16. Molecular Formula: NiCO3∙2Ni(OH)2∙XH2O. Color/form: green pwdr. Strem# 93-2808. http://www.strem.com/catalog/v/93-2808/
Catalog No. 50-901-14775