Learn More
Strem, An Ascensus Company CAS# 13478-00-7. 25g. Nickel(II) nitrate hexahydrate (99.9985%-Ni) PURATREM. MFCD00149805
SDP
Supplier:  Strem, An Ascensus Company 28144025G
Encompass
CAS# 13478-00-7. 25g. Nickel(II) nitrate hexahydrate (99.9985%-Ni) PURATREM. MFCD00149805. Molecular Weight: 182.72 (290.81). Molecular Formula: Ni(NO3)2∙6H2O. Color/form: green xtl. Strem# 28-1440. http://www.strem.com/catalog/v/28-1440/
Catalog No. 50-901-11258