Learn More
Gold Biotechnology Inc Nickel NTA Agarose Beads 50 mL
SDP
Supplier:  Gold Biotechnology Inc H35050
Encompass_Preferred
Nickel-NTA Agarose Resin has a binding capacity of 50 mg/ml In addition it allows for purification of proteins under native or denaturing conditions GoldBio Nickel Agarose works very well with the His-Tag Buffer Set
Catalog No. 50-005-62144