Learn More
Fine Science Tools Nickel Plated Pin Holder, 17 cm, each
SDP
Supplier:  Fine Science Tools 2601817
Encompass
Nickel Plated Pin Holder - 17cm/ Jaw Opening Diameter: 0 to 1mm/ Alloy: Nickel/Aluminum/ Length: 17cm/ Unit: Each
Catalog No. NC9316733