Learn More
Strem, An Ascensus Company CAS# 940895-79-4. 5g. Bis(N,N'-di-t-butylacetamidinato)nickel(II), (99.999%-Ni) PURATREM. MFCD28144557
SDP
Supplier:  Strem, An Ascensus Company 2800455G
Encompass
CAS# 940895-79-4. 5g. Bis(N,N'-di-t-butylacetamidinato)nickel(II), (99.999%-Ni) PURATREM. MFCD28144557. Molecular Weight: 397.27. Molecular Formula: C20H42N4Ni. Color/form: dark, purple-black xtl. Strem# 28-0045. http://www.strem.com/catalog/v/28-0045/
Catalog No. 50-204-5285