Learn More
Hach Company Nickel TNTplus plus Vial Test (0.1-6.0 mg/L Ni), 25 Tests
Supplier:  Hach Company TNT856
Encompass
Nickel TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nickel by the Dimethylglyoxime Method. Add Metals Prep Set (TNT890) to determine total nickel (requires digestion).
Catalog No. 50-199-8144