Learn More
Strem, An Ascensus Company CAS# 7440-02-0. 120cm. Nickel tubing (99.5%). MFCD00011137
SDP
Supplier:  Strem, An Ascensus Company 932876120CM
Encompass
CAS# 7440-02-0. 120cm. Nickel tubing (99.5%). MFCD00011137. Molecular Weight: 58.71. Molecular Formula: Ni. Color/form: 6.35 mm O.D. x 0.35 mm wall Strem# 93-2876. http://www.strem.com/catalog/v/93-2876/
Catalog No. 50-901-14846