Learn More
Sigma Aldrich Fine Chemicals Biosciences NickelII chloride hexahydr
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2233872KG
Encompass
Nickel(II) chloride hexahydrate is a hydrated nickel salt. Its crystals were monoclinc and crystal structure was investigated by X-ray single crystal method.
Catalog No. 50-289-9549