Learn More
Crescent Chemical Co Inc NICLOSAMIDE 100 MG
SDP
Supplier:  Crescent Chemical Co Inc PSC15510000
Encompass_Preferred
Niclosamid Pesticide other (WS) CAS 50-65-7 Molecular Weight - 327.1196 Storage - RT Catalog Num - DRE-C15510000 pesticide
Catalog No. 50-746-69