Learn More
Medchemexpress LLC HY-B0497 500mg Medchemexpress, Niclosamide CAS:50-65-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0497500MG
Encompass_Preferred
Medchemexpress, HY-B0497 500mg Niclosamide CAS:50-65-7 Niclosamide (BAY2353) is an orally bioavailable chlorinated salicylanilide, with anthelmintic and potential antineoplastic activity. Niclosamide (BAY2353) inhibits STAT3 with IC50 of 0.25 μM in HeLa cells and inhibits DNA replication in a cell-free assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9400