Learn More
Selleck Chemical LLC Niclosamide 50mg 50-65-7 Niclocide
SDP
Supplier:  Selleck Chemical LLC S303050MG
Encompass_Preferred
Niclosamide (Niclocide) can inhibit DNA replication and inhibit STAT3 with IC50 of 0.7 ?M in a cell-free assay. Niclosamide selectively inhibited the phosphorylation of STAT3 and had no obvious inhibition against the activation of other homologues (e.g., STAT1 and STAT5). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1332514