Learn More
Medchemexpress LLC HY-B0497 5g , Niclosamide CAS:50-65-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB04975G
Encompass_Preferred
HY-B0497 5g Niclosamide CAS: 50-65-7 Niclosamide (BAY2353) is an orally active antihelminthic agent used in parasitic infection research. Niclosamide is a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells[4]. Niclosamide has biological activities against cancer, inhibits DNA replication in Vero E6 cells[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9812