Learn More
Medchemexpress LLC Niclosamide (olamine) | 1420-04-8 | 98.66% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB0497C10G
Encompass_Preferred
Niclosamide olamine is an orally active antihelminthic agent used in parasitic infection research. It acts as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. This compound demonstrates biological activities against cancer and inhibits DNA replication in Vero E6 cells.
  • Orally active antihelminthic agent
  • Used in parasitic infection research
  • STAT3 inhibitor
  • Demonstrates biological activities against cancer
  • Inhibits DNA replication in Vero E6 cells
  • Inhibits adrenocortical carcinoma cellular proliferation
  • Suppresses colony formation and reduces invasive properties of endometrial cancer cells
  • Inhibits tumor growth in vivo
Catalog No. 50-004-07835